Learn More
1-Bromodecane, 98%
CAS: 112-29-8 | C10H21Br | 221.18 g/mol
Supplier: Thermo Scientific Chemicals A113400C
Description
1-Bromodecane is used to produce decanal. It is also used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromodecane is used to produce decanal. It is also used in organic synthesis.
Solubility
Miscible with most common organic solvents like ethanol, ethyl ether and chloroform. Not miscible in water.
Notes
Do not store together with strongly basic or oxidizing materials. Incompatible with strong oxidizing agents, strong bases.
Specifications
| 1-Bromodecane | |
| -30°C | |
| 238°C | |
| C10H21Br | |
| CH3(CH2)9Br | |
| 10,000 g | |
| 1735227 | |
| Miscible with most common organic solvents like ethanol,ethyl ether and chloroform. Not miscible in water. | |
| CCCCCCCCCCBr | |
| 221.18 | |
| 221.19 |
| 112-29-8 | |
| 1.067 | |
| 94°C (201°F) | |
| 1.456 | |
| MFCD00000221 | |
| UN2810 | |
| decyl bromide, n-decyl bromide, decane, 1-bromo, 1-decyl bromide, decylbromide, bromodecane, decane, bromo, labotest-bb ltbb001164, decylmonobromide, n-decylbromide | |
| MYMSJFSOOQERIO-UHFFFAOYSA-N | |
| 1-bromodecane | |
| 8173 | |
| 98% |
Chemical Identifiers
| 112-29-8 | |
| 221.18 | |
| MYMSJFSOOQERIO-UHFFFAOYSA-N | |
| 8173 | |
| CCCCCCCCCCBr |
| C10H21Br | |
| MFCD00000221 | |
| decyl bromide, n-decyl bromide, decane, 1-bromo, 1-decyl bromide, decylbromide, bromodecane, decane, bromo, labotest-bb ltbb001164, decylmonobromide, n-decylbromide | |
| 1-bromodecane |
Safety and Handling
GHS H Statement
H331
Toxic if inhaled.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H332-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 203-955-3
RTECSNumber : HD6850000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only