missing translation for 'onlineSavingsMsg'
Learn More

1-Bromodecane, 98%

Catalog No. AAA1134022
Change view
Click to view available options
Quantity:
10,000 g
100 g
500 g
2500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1134022 100 g
AAA1134036 500 g
AAA113400E 2500 g
AAA113400C 10,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1134022 Supplier Thermo Scientific Chemicals Supplier No. A1134022

CAS: 112-29-8 | C10H21Br | 221.18 g/mol

1-Bromodecane is used to produce decanal. It is also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromodecane is used to produce decanal. It is also used in organic synthesis.

Solubility
Miscible with most common organic solvents like ethanol, ethyl ether and chloroform. Not miscible in water.

Notes
Do not store together with strongly basic or oxidizing materials. Incompatible with strong oxidizing agents, strong bases.

Chemical Identifiers

CAS 112-29-8
Molecular Formula C10H21Br
Molecular Weight (g/mol) 221.18
MDL Number MFCD00000221
InChI Key MYMSJFSOOQERIO-UHFFFAOYSA-N
Synonym decyl bromide, n-decyl bromide, decane, 1-bromo, 1-decyl bromide, decylbromide, bromodecane, decane, bromo, labotest-bb ltbb001164, decylmonobromide, n-decylbromide
PubChem CID 8173
IUPAC Name 1-bromodecane
SMILES CCCCCCCCCCBr

Specifications

Melting Point -30°C
Density 1.067
Boiling Point 238°C
Flash Point 94°C (201°F)
Refractive Index 1.456
Linear Formula CH3(CH2)9Br
Quantity 100 g
UN Number UN2810
Beilstein 1735227
Solubility Information Miscible with most common organic solvents like ethanol,ethyl ether and chloroform. Not miscible in water.
Formula Weight 221.19
Percent Purity 98%
Chemical Name or Material 1-Bromodecane
Show More Show Less
Hazard Category H315-H319-H332-H335
Hazard Statement GHS H Statement
H331
Toxic if inhaled.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 203-955-3
RTECSNumber HD6850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only