missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoanthracene 97.0+%, TCI America™

Catalog No. B2615100MG
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2615100MG 100 mg
B2615500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2615100MG Supplier TCI America Supplier No. B2615100MG
Only null left

Chemical Identifiers

CAS 7397-92-4
Molecular Formula C14H9Br
Molecular Weight (g/mol) 257.13
MDL Number MFCD08276282
InChI Key XMWJLKOCNKJERQ-UHFFFAOYSA-N
PubChem CID 12529827
IUPAC Name 1-bromoanthracene
SMILES C1=CC=C2C=C3C(=CC2=C1)C=CC=C3Br

Specifications

Melting Point 100°C
Quantity 100 mg
Formula Weight 257.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromoanthracene