missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-6-chlorohexane 95.0+%, TCI America™

Catalog No. B1998500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1998500G 500 g
B199825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1998500G Supplier TCI America Supplier No. B1998500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6294-17-3
Molecular Formula C6H12BrCl
Molecular Weight (g/mol) 199.52
MDL Number MFCD00001019
InChI Key JTYUIAOHIYZBPB-UHFFFAOYSA-N
Synonym 6-chlorohexyl bromide, hexane, 1-bromo-6-chloro, 1-chloro-6-bromohexane, hexyl bromide, 6-chloro, 6-bromohexyl chloride, 1-bromo-6-chloro hexane, 1-bromo-6-chloro-hexane, acmc-1b9m0, 1-bromo-6-chlorohexane, jtyuiaohiyzbpb-uhfffaoysa
PubChem CID 80516
IUPAC Name 1-bromo-6-chlorohexane
SMILES ClCCCCCCBr

Specifications

Color Colorless
Boiling Point 96°C
Quantity 500 g
Formula Weight 199.52
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-6-chlorohexane
EINECSNumber (2)-3316
TSCA Yes