accessibility menu, dialog, popup

UserName

ClBr

CAS RN 6294-17-3

IUPAC Name: 1-bromo-6-chlorohexane
Synonyms: 1-bromo-6-chlorohexane
Molecular Weight (g/mol): 199.52
Molecular Formula: C6H12BrCl
InChi Key: JTYUIAOHIYZBPB-UHFFFAOYSA-N
SMILES: ClCCCCCCBr
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)