missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-5-phenylpentane, 98%

CAS: 14469-83-1 | C11H15Br | 227.14 g/mol

$521.17 - $521.17

Chemical Identifiers

CAS 14469-83-1
Molecular Formula C11H15Br
Molecular Weight (g/mol) 227.14
MDL Number MFCD01075177
InChI Key QICUPOFVENZWSC-UHFFFAOYSA-N
Synonym 5-bromopentyl benzene, 1-bromo-5-phenylpentane, benzene, 5-bromopentyl, 5-phenylpentyl bromide, 1-bromo-5-phenyl-pentane, 5-bromo-n-pentyl benzene, 5-bromo-pentyl-benzene, 5-brompentyl benzol, phenylpentyl bromide, 5-phenylpentylbromide
PubChem CID 285561
IUPAC Name 5-bromopentylbenzene
SMILES C1=CC=C(C=C1)CCCCCBr
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC384740050
SDS View Documents
Thermo Scientific Chemicals
384740050
5 g Glass bottle
Each for $521.17
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14469-83-1
227.14
QICUPOFVENZWSC-UHFFFAOYSA-N
285561
C1=CC=C(C=C1)CCCCCBr
C11H15Br
MFCD01075177
5-bromopentyl benzene, 1-bromo-5-phenylpentane, benzene, 5-bromopentyl, 5-phenylpentyl bromide, 1-bromo-5-phenyl-pentane, 5-bromo-n-pentyl benzene, 5-bromo-pentyl-benzene, 5-brompentyl benzol, phenylpentyl bromide, 5-phenylpentylbromide
5-bromopentylbenzene

Specifications

14469-83-1
100.0
1.2300g/mL
Authentic
Glass bottle
1.5320 to 1.534
MFCD01075177
1.23
QICUPOFVENZWSC-UHFFFAOYSA-N
5-bromopentylbenzene
285561
98%
1-Bromo-5-phenylpentane
97.5
Colorless
148°C (18.0 mmHg)
97.5% min. (GC)
C11H15Br
C6H5(CH2)5Br
5 g
5-bromopentyl benzene, 1-bromo-5-phenylpentane, benzene, 5-bromopentyl, 5-phenylpentyl bromide, 1-bromo-5-phenyl-pentane, 5-bromo-n-pentyl benzene, 5-bromo-pentyl-benzene, 5-brompentyl benzol, phenylpentyl bromide, 5-phenylpentylbromide
C1=CC=C(C=C1)CCCCCBr
227.14
227.14
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only