missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-4-(trimethylsilyl)benzene 97.0+%, TCI America™
$203.17 - $725.11
Chemical Identifiers
| CAS | 6999-03-7 |
|---|---|
| Molecular Formula | C9H13BrSi |
| Molecular Weight (g/mol) | 229.19 |
| MDL Number | MFCD00094165 |
| InChI Key | UKTSSJJZFVGTCG-UHFFFAOYSA-N |
| Synonym | (4-Bromophenyl)trimethylsilane |
| PubChem CID | 138897 |
| IUPAC Name | (4-bromophenyl)-trimethylsilane |
| SMILES | C[Si](C)(C)C1=CC=C(C=C1)Br |
Chemical Identifiers
| 6999-03-7 | |
| 229.19 | |
| UKTSSJJZFVGTCG-UHFFFAOYSA-N | |
| 138897 | |
| C[Si](C)(C)C1=CC=C(C=C1)Br |
| C9H13BrSi | |
| MFCD00094165 | |
| (4-Bromophenyl)trimethylsilane | |
| (4-bromophenyl)-trimethylsilane |
Specifications
| 6999-03-7 | |
| 230°C | |
| MFCD00094165 | |
| (4-Bromophenyl)trimethylsilane | |
| C[Si](C)(C)C1=CC=C(C=C1)Br | |
| 229.19 | |
| 229.19 | |
| Liquid |
| Yellow | |
| C9H13BrSi | |
| 5 g | |
| UKTSSJJZFVGTCG-UHFFFAOYSA-N | |
| (4-bromophenyl)-trimethylsilane | |
| 138897 | |
| ≥97.0% (GC) | |
| 1-Bromo-4-(trimethylsilyl)benzene |
Safety and Handling
TSCA : No