missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-(trifluoromethylthio)benzene, 97%

Catalog No. AAB2087403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2087403 1 g
AAB2087406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2087403 Supplier Thermo Scientific Chemicals Supplier No. B2087403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 333-47-1 | C7H4BrF3S | 257.068 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 333-47-1
Molecular Formula C7H4BrF3S
Molecular Weight (g/mol) 257.068
MDL Number MFCD00040840
InChI Key CQQBVMXLTBXNLB-UHFFFAOYSA-N
Synonym 4-trifluoromethylthio bromobenzene, 1-bromo-4-trifluoromethyl sulfanyl benzene, 4-bromophenyl trifluoromethyl sulfide, 4-bromophenyl trifluoromethyl sulphide, 1-bromo-4-trifluoromethylthio benzene, 1-bromo-4-trifluoromethylthiobenzene, 4-bromophenyl trifluoromethyl sulfane, 1-bromo-4-trifluoromethyl thio benzene, 4-bromo-1-trifluoromethylthio benzene, 1-bromo-4-trifluoromethylsulfanyl benzene
PubChem CID 736413
IUPAC Name 1-bromo-4-(trifluoromethylsulfanyl)benzene
SMILES C1=CC(=CC=C1SC(F)(F)F)Br

Specifications

Density 1.71
Boiling Point 199°C to 200°C
Flash Point 83°C (181°F)
Refractive Index 1.517
Quantity 1 g
Beilstein 2361926
Formula Weight 257.07
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-(trifluoromethylthio)benzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only