missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-(trifluoromethoxy)benzene 98.0+%, TCI America™

Catalog No. B17725G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17725G 5 g
B177225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17725G Supplier TCI America Supplier No. B17725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 407-14-7
Molecular Formula C7H4BrF3O
Molecular Weight (g/mol) 241.01
MDL Number MFCD00040834
InChI Key SEAOBYFQWJFORM-UHFFFAOYSA-N
Synonym 1-bromo-4-trifluoromethoxy benzene, 4-trifluoromethoxy bromobenzene, 4-bromo trifluoromethoxy benzene, 4-trifluoromethoxybromobenzene, p-bromotrifluoromethoxybenzene, benzene, 1-bromo-4-trifluoromethoxy, 4-bromo-1-trifluoromethoxy benzene, p-trifluoromethoxybromobenzene, 4-bromotrifluoromethoxybenzene, 4-bromo-trifluoromethoxybenzene
PubChem CID 521008
IUPAC Name 1-bromo-4-(trifluoromethoxy)benzene
SMILES FC(F)(F)OC1=CC=C(Br)C=C1

Specifications

Color Yellow
Boiling Point 80°C
Quantity 5 g
Formula Weight 241.01
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-(trifluoromethoxy)benzene