missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-(trifluoromethoxy)benzene, 98%

Catalog No. AAA1210414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1210414 25 g
AAA1210406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1210414 Supplier Thermo Scientific Chemicals Supplier No. A1210414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 407-14-7 | C7H4BrF3O | 241.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 407-14-7
Molecular Formula C7H4BrF3O
Molecular Weight (g/mol) 241.01
MDL Number MFCD00040834
InChI Key SEAOBYFQWJFORM-UHFFFAOYSA-N
Synonym 1-bromo-4-trifluoromethoxy benzene, 4-trifluoromethoxy bromobenzene, 4-bromo trifluoromethoxy benzene, 4-trifluoromethoxybromobenzene, p-bromotrifluoromethoxybenzene, benzene, 1-bromo-4-trifluoromethoxy, 4-bromo-1-trifluoromethoxy benzene, p-trifluoromethoxybromobenzene, 4-bromotrifluoromethoxybenzene, 4-bromo-trifluoromethoxybenzene
PubChem CID 521008
IUPAC Name 1-bromo-4-(trifluoromethoxy)benzene
SMILES FC(F)(F)OC1=CC=C(Br)C=C1

Specifications

Density 1.622
Boiling Point 155°C to 157°C
Flash Point 67°C (152°F)
Odor Unpleasant
Refractive Index 1.461
Quantity 25 g
Beilstein 2046332
Formula Weight 241.01
Percent Purity 98%
Chemical Name or Material 1-Bromo-4- (trifluoromethoxy)benzene
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 206-979-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only