missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-4-tert-butylbenzene 96.0+%, TCI America™
$68.97 - $197.59
Chemical Identifiers
| CAS | 3972-65-4 |
|---|---|
| Molecular Formula | C10H13Br |
| Molecular Weight (g/mol) | 213.118 |
| MDL Number | MFCD00000108 |
| InChI Key | XHCAGOVGSDHHNP-UHFFFAOYSA-N |
| Synonym | 4-tert-butylbromobenzene, 1-bromo-4-tert-butyl benzene, butylbromobenzene, p-bromo-tert-butylbenzene, 1-bromo-4-t-butylbenzene, 1-bromo-4-1,1-dimethylethyl benzene, 4-tert-butyl-1-bromobenzene, p-bromo-t-butylbenzene, 1-bromo-p-t-butylbenzene, 4-bromo-tert-butylbenzene |
| PubChem CID | 77595 |
| IUPAC Name | 1-bromo-4-tert-butylbenzene |
| SMILES | CC(C)(C)C1=CC=C(C=C1)Br |
Chemical Identifiers
| 3972-65-4 | |
| 213.118 | |
| XHCAGOVGSDHHNP-UHFFFAOYSA-N | |
| 77595 | |
| CC(C)(C)C1=CC=C(C=C1)Br |
| C10H13Br | |
| MFCD00000108 | |
| 4-tert-butylbromobenzene, 1-bromo-4-tert-butyl benzene, butylbromobenzene, p-bromo-tert-butylbenzene, 1-bromo-4-t-butylbenzene, 1-bromo-4-1,1-dimethylethyl benzene, 4-tert-butyl-1-bromobenzene, p-bromo-t-butylbenzene, 1-bromo-p-t-butylbenzene, 4-bromo-tert-butylbenzene | |
| 1-bromo-4-tert-butylbenzene |
Specifications
| 3972-65-4 | |
| 228°C | |
| MFCD00000108 | |
| 4-tert-butylbromobenzene, 1-bromo-4-tert-butyl benzene, butylbromobenzene, p-bromo-tert-butylbenzene, 1-bromo-4-t-butylbenzene, 1-bromo-4-1,1-dimethylethyl benzene, 4-tert-butyl-1-bromobenzene, p-bromo-t-butylbenzene, 1-bromo-p-t-butylbenzene, 4-bromo-tert-butylbenzene | |
| CC(C)(C)C1=CC=C(C=C1)Br | |
| 213.118 | |
| 213.12 | |
| Liquid |
| Yellow | |
| C10H13Br | |
| 25 g | |
| XHCAGOVGSDHHNP-UHFFFAOYSA-N | |
| 1-bromo-4-tert-butylbenzene | |
| 77595 | |
| ≥96.0% (GC) | |
| 1-Bromo-4-tert-butylbenzene |
Safety and Handling
TSCA : Yes