missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-tert-butylbenzene 96.0+%, TCI America™

Catalog No. B172725G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B172725G 25 g
B1727100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B172725G Supplier TCI America Supplier No. B172725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3972-65-4
Molecular Formula C10H13Br
Molecular Weight (g/mol) 213.118
MDL Number MFCD00000108
InChI Key XHCAGOVGSDHHNP-UHFFFAOYSA-N
Synonym 4-tert-butylbromobenzene, 1-bromo-4-tert-butyl benzene, butylbromobenzene, p-bromo-tert-butylbenzene, 1-bromo-4-t-butylbenzene, 1-bromo-4-1,1-dimethylethyl benzene, 4-tert-butyl-1-bromobenzene, p-bromo-t-butylbenzene, 1-bromo-p-t-butylbenzene, 4-bromo-tert-butylbenzene
PubChem CID 77595
IUPAC Name 1-bromo-4-tert-butylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)Br

Specifications

Color Yellow
Boiling Point 228°C
Quantity 25 g
Formula Weight 213.12
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-tert-butylbenzene