missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-tert-butylbenzene, 97%

Catalog No. p-7022933
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
AAA1592414 25 g
AAA1592422 100 g
AAA1592436 500 g
3 options

Catalog No. AAA1592414

Supplier: Thermo Scientific Chemicals A1592414

Only null left

CAS: 3972-65-4 | C10H13Br | 213.118 g/mol

1-Bromo-4-tert-butylbenzene was used in the synthesis of 4-tert-butyl-phenylboronic acid1, 1-deoxy analogs of CP-47,497 (n = 0 to 7) and 1-deoxy analogs of CP-55,940 (n = 0 to 7). It undergoes lithium-bromide exchange reactions with n-butyllithium and tert-butyllithium at 0°C in various solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-tert-butylbenzene was used in the synthesis of 4-tert-butyl-phenylboronic acid1, 1-deoxy analogs of CP-47,497 (n = 0 to 7) and 1-deoxy analogs of CP-55,940 (n = 0 to 7). It undergoes lithium-bromide exchange reactions with n-butyllithium and tert-butyllithium at 0°C in various solvents.

Solubility
Insoluble in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 3972-65-4
Molecular Formula C10H13Br
Molecular Weight (g/mol) 213.118
MDL Number MFCD00000108
InChI Key XHCAGOVGSDHHNP-UHFFFAOYSA-N
Synonym 4-tert-butylbromobenzene, 1-bromo-4-tert-butyl benzene, butylbromobenzene, p-bromo-tert-butylbenzene, 1-bromo-4-t-butylbenzene, 1-bromo-4-1,1-dimethylethyl benzene, 4-tert-butyl-1-bromobenzene, p-bromo-t-butylbenzene, 1-bromo-p-t-butylbenzene, 4-bromo-tert-butylbenzene
PubChem CID 77595
IUPAC Name 1-bromo-4-tert-butylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)Br

Specifications

Melting Point 15°C to 16°C
Density 1.229
Boiling Point 231°C to 232°C
Flash Point 97°C (206°F)
Refractive Index 1.533
Quantity 25 g
Beilstein 1859117
Solubility Information Insoluble in water.
Formula Weight 213.12
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-tert-butylbenzene
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 223-599-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only