missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-tert-butylbenzene, 97%

Catalog No. AC225190250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC225190250 25 g Glass bottle
AC225191000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC225190250 Supplier Thermo Scientific Chemicals Supplier No. 225190250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3972-65-4 | C10H13Br | 213.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3972-65-4
Molecular Formula C10H13Br
Molecular Weight (g/mol) 213.12
MDL Number MFCD00000108
InChI Key XHCAGOVGSDHHNP-UHFFFAOYSA-N
Synonym 4-tert-butylbromobenzene, 1-bromo-4-tert-butyl benzene, butylbromobenzene, p-bromo-tert-butylbenzene, 1-bromo-4-t-butylbenzene, 1-bromo-4-1,1-dimethylethyl benzene, 4-tert-butyl-1-bromobenzene, p-bromo-t-butylbenzene, 1-bromo-p-t-butylbenzene, 4-bromo-tert-butylbenzene
PubChem CID 77595
IUPAC Name 1-bromo-4-tert-butylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 15°C to 16°C
Color Colorless to Yellow
Density 1.2200g/mL
Boiling Point 80°C to 81°C (2.0 mmHg)
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5315 to 1.5345
Linear Formula (CH3)3CC6H4Br
Quantity 25 g
Beilstein 05, 416
Specific Gravity 1.22
Formula Weight 213.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-tert-butylbenzene, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 223-599-2
TSCA TSCA

RUO – Research Use Only