missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-4-propylbenzene, TCI America™

$143.61 - $487.38

Chemical Identifiers

CAS 588-93-2
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.091
MDL Number MFCD00012456
InChI Key NUPWGLKBGVNSJX-UHFFFAOYSA-N
Synonym 1-bromo-4-n-propylbenzene, 4-bromo-n-propylbenzene, 1-4'-bromophenyl propane, 1-bromo-4-propyl-benzene, benzene, 1-bromo-4-propyl, 4-propylbromobenzene, 4-propyl bromobenzene, 4-n-propylbromobenzene, 4-propylphenyl bromide, pubchem10762
PubChem CID 136374
IUPAC Name 1-bromo-4-propylbenzene
SMILES CCCC1=CC=C(C=C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B18895G View Documents TCI America
B18895G
5g Each for $143.61
 
B188925G View Documents TCI America
B188925G
25g Each for $487.38
 
Specifications

Chemical Identifiers

588-93-2
199.091
NUPWGLKBGVNSJX-UHFFFAOYSA-N
136374
CCCC1=CC=C(C=C1)Br
C9H11Br
MFCD00012456
1-bromo-4-n-propylbenzene, 4-bromo-n-propylbenzene, 1-4'-bromophenyl propane, 1-bromo-4-propyl-benzene, benzene, 1-bromo-4-propyl, 4-propylbromobenzene, 4-propyl bromobenzene, 4-n-propylbromobenzene, 4-propylphenyl bromide, pubchem10762
1-bromo-4-propylbenzene

Specifications

225°C
5g
C9H11Br
1-bromo-4-n-propylbenzene, 4-bromo-n-propylbenzene, 1-4'-bromophenyl propane, 1-bromo-4-propyl-benzene, benzene, 1-bromo-4-propyl, 4-propylbromobenzene, 4-propyl bromobenzene, 4-n-propylbromobenzene, 4-propylphenyl bromide, pubchem10762
CCCC1=CC=C(C=C1)Br
199.091
199.09
≥97.0% (GC)
Yellow
588-93-2
MFCD00012456
NUPWGLKBGVNSJX-UHFFFAOYSA-N
1-bromo-4-propylbenzene
136374
Clear Liquid at 20°C
1-Bromo-4-propylbenzene
Safety and Handling

Safety and Handling

TSCA : No