missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-propylbenzene 97.0+%, TCI America™

Catalog No. B18895G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18895G 5 g
B188925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18895G Supplier TCI America Supplier No. B18895G

Chemical Identifiers

CAS 588-93-2
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.091
MDL Number MFCD00012456
InChI Key NUPWGLKBGVNSJX-UHFFFAOYSA-N
Synonym 1-bromo-4-n-propylbenzene, 4-bromo-n-propylbenzene, 1-4'-bromophenyl propane, 1-bromo-4-propyl-benzene, benzene, 1-bromo-4-propyl, 4-propylbromobenzene, 4-propyl bromobenzene, 4-n-propylbromobenzene, 4-propylphenyl bromide, pubchem10762
PubChem CID 136374
IUPAC Name 1-bromo-4-propylbenzene
SMILES CCCC1=CC=C(C=C1)Br

Specifications

Color Yellow
Boiling Point 225°C
Quantity 5 g
Formula Weight 199.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-propylbenzene