missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-(phenoxymethyl)benzene, 97%, Thermo Scientific™

Catalog No. CC63710CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC63710CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC63710CB Supplier Thermo Scientific Chemicals Supplier No. CC63710CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20600-22-0
Molecular Formula C13H11BrO
Molecular Weight (g/mol) 263.134
MDL Number MFCD00017847
InChI Key KZVLWVAXLYUCLW-UHFFFAOYSA-N
Synonym 1-bromo-4-phenoxymethyl benzene, 4-bromophenyl methoxy benzene, 4-bromobenzylphenyl ether, 4-bromobenzyl phenyl ether, 1-bromo4-phenoxymethyl-benzene, 1-bromo-4-phenoxymethyl-benzene, benzyl phenyl ether analogue, 6c
PubChem CID 726149
IUPAC Name 1-bromo-4-(phenoxymethyl)benzene
SMILES C1=CC=C(C=C1)OCC2=CC=C(C=C2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 263.13
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-(phenoxymethyl)benzene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.