missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-nitrobenzene 99.0+%, TCI America™

Catalog No. p-7118570
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B062425G 25 g
B0624250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B062425G Supplier TCI America Supplier No. B062425G
Only null left

Chemical Identifiers

CAS 586-78-7
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.01
MDL Number MFCD00007280
InChI Key ZDFBKZUDCQQKAC-UHFFFAOYSA-N
Synonym 4-bromonitrobenzene, 4-nitrobromobenzene, p-bromonitrobenzene, benzene, 1-bromo-4-nitro, p-nitrobromobenzene, 4-nitrophenyl bromide, 4-bromo-1-nitrobenzene, p-nitrophenyl bromide, 1-bromo-4-nitro-benzene, ccris 3115
PubChem CID 11466
IUPAC Name 1-bromo-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 127°C
Boiling Point 256°C
Quantity 25 g
UN Number 3459
Formula Weight 202.01
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-4-nitrobenzene
RTECSNumber CY9040550
TSCA Yes