missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-nitrobenzene, 98%

Catalog No. p-7023799
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1770506 5 g
AAA1770514 25 g
AAA1770522 100 g
AAA1770536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1770506 Supplier Thermo Scientific Chemicals Supplier No. A1770506
Only null left

CAS: 586-78-7 | C6H4BrNO2 | 202.01 g/mol

It can be employed as a syntheses material intermediate. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be employed as a syntheses material intermediate. It is also used as a pharmaceutical intermediate.

Solubility
Insoluble in water. Almost transparent in hot ethyl alcohol. Soluble in ether

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed in a cool, dry place.

Chemical Identifiers

CAS 586-78-7
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.01
MDL Number MFCD00007280
InChI Key ZDFBKZUDCQQKAC-UHFFFAOYSA-N
Synonym 4-bromonitrobenzene, 4-nitrobromobenzene, p-bromonitrobenzene, benzene, 1-bromo-4-nitro, p-nitrobromobenzene, 4-nitrophenyl bromide, 4-bromo-1-nitrobenzene, p-nitrophenyl bromide, 1-bromo-4-nitro-benzene, ccris 3115
PubChem CID 11466
IUPAC Name 1-bromo-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 124°C to 127°C
Density 1.948
Boiling Point 254°C to 256°C
Quantity 5 g
UN Number UN3459
Beilstein 636964
Solubility Information Insoluble in water. Almost transparent in hot ethyl alcohol. Soluble in ether
Formula Weight 202.01
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-nitrobenzene
Hazard Category H302+H312+H332-H315-H319-H335-H373
Hazard Statement GHS H Statement
H331-H373-H302-H312-H315-H319-H335
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROBROMOBENZENES, SOLID
EINECSNumber 209-583-8
RTECSNumber CY9040550
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only