missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-n-propylbenzene, 99%

Catalog No. AAH2623622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2623622 100 g
AAH2623606 5 g
AAH2623614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2623622 Supplier Thermo Scientific Chemicals Supplier No. H2623622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 588-93-2 | C9H11Br | 199.091 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 588-93-2
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.091
MDL Number MFCD00012456
InChI Key NUPWGLKBGVNSJX-UHFFFAOYSA-N
Synonym 1-bromo-4-n-propylbenzene, 4-bromo-n-propylbenzene, 1-4'-bromophenyl propane, 1-bromo-4-propyl-benzene, benzene, 1-bromo-4-propyl, 4-propylbromobenzene, 4-propyl bromobenzene, 4-n-propylbromobenzene, 4-propylphenyl bromide, pubchem10762
PubChem CID 136374
IUPAC Name 1-bromo-4-propylbenzene
SMILES CCCC1=CC=C(C=C1)Br

Specifications

Density 1.286
Boiling Point 225°C
Flash Point 95°C (203°F)
Refractive Index 1.537
Quantity 100 g
Formula Weight 199.09
Percent Purity 99%
Chemical Name or Material 1-Bromo-4-n-propylbenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only