missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-(n-octyloxy)benzene, 98%

Catalog No. AAA1333403
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1333403 1 g
AAA1333414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1333403 Supplier Thermo Scientific Chemicals Supplier No. A1333403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 96693-05-9 | C14H21BrO | 285.225 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96693-05-9
Molecular Formula C14H21BrO
Molecular Weight (g/mol) 285.225
MDL Number MFCD00043445
InChI Key UVBFFPZGOOKWNR-UHFFFAOYSA-N
Synonym p-bromophenyl octyl ether, 1-bromo-4-octyloxy benzene, 1-bromo-4-octyloxybenzene, 4-n-octyloxybromobenzene, 1-bromo-4-n-octyloxy benzene, 4-bromo-1-octyloxybenzene, 4-octyloxybromobenzene, 4-bromo-octyloxybenzene, 4-bromophenyloctyl ether, 1-4-bromophenoxy octane
PubChem CID 145298
IUPAC Name 1-bromo-4-octoxybenzene
SMILES CCCCCCCCOC1=CC=C(C=C1)Br

Specifications

Boiling Point 135°C to 137°C (0.5 mmHg)
Refractive Index 1.518
Quantity 1 g
Formula Weight 285.23
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-(n-octyloxy)benzene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only