missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-n-octylbenzene 98.0+%, TCI America™

Catalog No. p-7119493
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21175G 5 g
B211725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21175G Supplier TCI America Supplier No. B21175G

Chemical Identifiers

CAS 51554-93-9
Molecular Formula C14H21Br
Molecular Weight (g/mol) 269.226
MDL Number MFCD00038348
InChI Key OOZQSVXPBCINJF-UHFFFAOYSA-N
Synonym 1-4-bromophenyl octane, 1-bromo-4-n-octylbenzene, p-octylbromobenzene, p-bromophenyloctane, 4-n-octylbromobenzene, 1-p-bromophenyl octane, 4-bromooctylbenzene, 4-oct-1-yl bromobenzene, benzene, 1-bromo-4-octyl, 4-bromo-1-octylbenzene
PubChem CID 142854
IUPAC Name 1-bromo-4-octylbenzene
SMILES CCCCCCCCC1=CC=C(C=C1)Br

Specifications

Color Yellow
Boiling Point 125°C
Quantity 5 g
Formula Weight 269.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-n-octylbenzene