missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-n-octylbenzene 90.0+%, TCI America™

Catalog No. p-7119155
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
B16105G 5 g
B161025G 25 g
2 options

Catalog No. B16105G

Supplier: TCI America B16105G

Only null left

Chemical Identifiers

CAS 51554-93-9
Molecular Formula C14H21Br
Molecular Weight (g/mol) 269.226
MDL Number MFCD00038348
InChI Key OOZQSVXPBCINJF-UHFFFAOYSA-N
Synonym 1-4-bromophenyl octane, 1-bromo-4-n-octylbenzene, p-octylbromobenzene, p-bromophenyloctane, 4-n-octylbromobenzene, 1-p-bromophenyl octane, 4-bromooctylbenzene, 4-oct-1-yl bromobenzene, benzene, 1-bromo-4-octyl, 4-bromo-1-octylbenzene
PubChem CID 142854
IUPAC Name 1-bromo-4-octylbenzene
SMILES CCCCCCCCC1=CC=C(C=C1)Br

Specifications

Color Yellow
Boiling Point 148°C
Quantity 5 g
Formula Weight 269.23
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-n-octylbenzene