missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-n-hexyloxybenzene, 97%, Thermo Scientific Chemicals

Catalog No. p-7024848
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2008506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2008506 Supplier Thermo Scientific Chemicals Supplier No. B2008506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 30752-19-3 | Molecular Formula: C12H17BrO | Molecular Weight: 257.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30752-19-3
Molecular Formula C12H17BrO
Molecular Weight (g/mol) 257.171
MDL Number MFCD00173753
InChI Key GKLMJONYGGTHHM-UHFFFAOYSA-N
Synonym 1-bromo-4-hexyloxy benzene, 1-bromo-4-hexyloxybenzene, 1-4-bromophenoxy hexane, 1-bromo-4-n-hexyloxybenzene, 4-n-hexyloxybromobenzene, p-bromophenylhexyl ether, benzene, 1-bromo-4-hexyloxy, p-hexyloxybromobenzene, 4-hexyloxybromobenzene, 4-bromophenyl hexyl ether
PubChem CID 520492
IUPAC Name 1-bromo-4-hexoxybenzene
SMILES CCCCCCOC1=CC=C(C=C1)Br

Specifications

Flash Point >110°C (230°F)
Refractive Index 1.524
Quantity 5 g
Formula Weight 257.18
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-n-hexyloxybenzene
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only