missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-n-hexylbenzene, 97%

Catalog No. AAA1591806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1591806 5 g
AAA1591814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1591806 Supplier Thermo Scientific Chemicals Supplier No. A1591806

CAS: 23703-22-2 | C12H17Br | 241.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23703-22-2
Molecular Formula C12H17Br
Molecular Weight (g/mol) 241.17
MDL Number MFCD00061114
InChI Key MXHOLIARBWJKCR-UHFFFAOYSA-N
Synonym 1-4-bromophenyl hexane, 4-hexylbromobenzene, 1-bromo-4-n-hexylbenzene, 4-bromo-1-hexylbenzene, benzene, 1-bromo-4-hexyl, 4-bromo-n-hexylbenzene, 4-hexylphenyl bromide, 1-bromo-4-hexyl-benzene, acmc-1cb1g, #
PubChem CID 598186
IUPAC Name 1-bromo-4-hexylbenzene
SMILES CCCCCCC1=CC=C(Br)C=C1

Specifications

Density 1.179
Boiling Point 280°C
Flash Point >120°C (248°F)
Refractive Index 1.5213
Quantity 5 g
Formula Weight 241.17
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-n-hexylbenzene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only