missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-iodobenzene, 98%

Catalog No. p-4510030
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC218380250 25 g Glass bottle
AC218381000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC218380250 Supplier Thermo Scientific Chemicals Supplier No. 218380250
Only null left

CAS: 589-87-7 | C6H4BrI | 282.9 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 589-87-7
Molecular Formula C6H4BrI
Molecular Weight (g/mol) 282.9
InChI Key UCCUXODGPMAHRL-UHFFFAOYSA-N
Synonym 4-bromoiodobenzene, benzene, 1-bromo-4-iodo, p-bromoiodobenzene, p-iodobromobenzene, p-bromophenyl iodide, 4-bromo-1-iodobenzene, 1-bromo-4-iodo-benzene, 4-iodobromobenzene, rarechem fh 1g 0a12, p-bromo iodobenzene
PubChem CID 11522
IUPAC Name 1-bromo-4-iodobenzene
SMILES C1=CC(=CC=C1Br)I

Specifications

Melting Point 89.0°C to 93.0°C
Color White to Brown
Boiling Point 120.0°C to 122.0°C (14.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4I
Quantity 25 g
Beilstein 05,223
Formula Weight 282.9
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Needles
Chemical Name or Material 1-Bromo-4-iodobenzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 209-662-7

RUO – Research Use Only