missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-4-heptylbenzene (stabilized with Copper chip) 96.0+%, TCI America™
$44.73 - $130.48
Chemical Identifiers
| CAS | 76287-49-5 |
|---|---|
| Molecular Formula | C13H19Br |
| Molecular Weight (g/mol) | 255.199 |
| MDL Number | MFCD00061115 |
| InChI Key | YHKMTIJHJWYYAG-UHFFFAOYSA-N |
| Synonym | 1-bromo-4-n-heptylbenzene, 1-4-bromophenyl heptane, 4-heptylbromobenzene, benzene, 1-bromo-4-heptyl, 4-bromo-1-heptylbenzene, 1-bromo-4-heptyl-benzene, acmc-209p2u, 1-bromanyl-4-heptyl-benzene, ksc912e9b |
| PubChem CID | 2775100 |
| IUPAC Name | 1-bromo-4-heptylbenzene |
| SMILES | CCCCCCCC1=CC=C(C=C1)Br |
Chemical Identifiers
| 76287-49-5 | |
| 255.199 | |
| YHKMTIJHJWYYAG-UHFFFAOYSA-N | |
| 2775100 | |
| CCCCCCCC1=CC=C(C=C1)Br |
| C13H19Br | |
| MFCD00061115 | |
| 1-bromo-4-n-heptylbenzene, 1-4-bromophenyl heptane, 4-heptylbromobenzene, benzene, 1-bromo-4-heptyl, 4-bromo-1-heptylbenzene, 1-bromo-4-heptyl-benzene, acmc-209p2u, 1-bromanyl-4-heptyl-benzene, ksc912e9b | |
| 1-bromo-4-heptylbenzene |
Specifications
| 76287-49-5 | |
| 190°C | |
| MFCD00061115 | |
| 1-bromo-4-n-heptylbenzene, 1-4-bromophenyl heptane, 4-heptylbromobenzene, benzene, 1-bromo-4-heptyl, 4-bromo-1-heptylbenzene, 1-bromo-4-heptyl-benzene, acmc-209p2u, 1-bromanyl-4-heptyl-benzene, ksc912e9b | |
| CCCCCCCC1=CC=C(C=C1)Br | |
| 255.199 | |
| 255.20 | |
| Liquid |
| Yellow | |
| C13H19Br | |
| 1 g | |
| YHKMTIJHJWYYAG-UHFFFAOYSA-N | |
| 1-bromo-4-heptylbenzene | |
| 2775100 | |
| ≥96.0% (GC) | |
| 1-Bromo-4-heptylbenzene (stabilized with Copper chip) |
Safety and Handling
TSCA : No