missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-heptylbenzene (stabilized with Copper chip) 96.0+%, TCI America™

Catalog No. B16085G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16085G 5 g
B16081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16085G Supplier TCI America Supplier No. B16085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76287-49-5
Molecular Formula C13H19Br
Molecular Weight (g/mol) 255.199
MDL Number MFCD00061115
InChI Key YHKMTIJHJWYYAG-UHFFFAOYSA-N
Synonym 1-bromo-4-n-heptylbenzene, 1-4-bromophenyl heptane, 4-heptylbromobenzene, benzene, 1-bromo-4-heptyl, 4-bromo-1-heptylbenzene, 1-bromo-4-heptyl-benzene, acmc-209p2u, 1-bromanyl-4-heptyl-benzene, ksc912e9b
PubChem CID 2775100
IUPAC Name 1-bromo-4-heptylbenzene
SMILES CCCCCCCC1=CC=C(C=C1)Br

Specifications

Color Yellow
Boiling Point 190°C
Quantity 5 g
Formula Weight 255.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-heptylbenzene (stabilized with Copper chip)