missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-fluoronaphthalene 97.0+%, TCI America™

Catalog No. p-7123172
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37971G 1 g
B37975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37971G Supplier TCI America Supplier No. B37971G

Chemical Identifiers

CAS 341-41-3
Molecular Formula C10H6BrF
Molecular Weight (g/mol) 225.06
MDL Number MFCD00051473
InChI Key VAUJZKBFENPOCH-UHFFFAOYSA-N
Synonym 4-bromo-1-fluoronaphthalene, 1-fluoro-4-bromonaphthalene, naphthalene, 1-bromo-4-fluoro, acmc-1cnye, 1-bromo 4-fluoronaphthalene, 1-bromo-4-fluoro naphthalene, 1-bromo-4-fluoro-naphthalene, ksc493c4t, 1-bromo-4-fluoronaphthalene
PubChem CID 67647
IUPAC Name 1-bromo-4-fluoronaphthalene
SMILES C1=CC=C2C(=C1)C(=CC=C2Br)F

Specifications

Melting Point 37°C
Color White-Yellow
Quantity 1 g
Formula Weight 225.06
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Bromo-4-fluoronaphthalene
TSCA No
Recommended Storage Refrigerator