accessibility menu, dialog, popup

UserName

CAS RN 341-41-3

IUPAC Name: 1-bromo-4-fluoronaphthalene
Synonyms: 1-bromo-4-fluoronaphthalene
Molecular Weight (g/mol): 225.06
Molecular Formula: C10H6BrF
InChi Key: VAUJZKBFENPOCH-UHFFFAOYSA-N
SMILES: FC1=C2C=CC=CC2=C(Br)C=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)