missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-4-fluorobenzene, 99%
Description
1-Bromo-4-fluorobenzene is used as an intermediate in the synthesis of atypical antipsychotic agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromo-4-fluorobenzene is used as an intermediate in the synthesis of atypical antipsychotic agents.
Solubility
Soluble in water(0.14g/L).
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
1-Bromo-4-fluorobenzene is used as an intermediate in the synthesis of atypical antipsychotic agents.
Solubility
Soluble in water(0.14g/L).
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | -16°C |
| Density | 1.604 |
| Boiling Point | 151°C to 153°C |
| Flash Point | 60°C (140°F) |
| Odor | Aromatic |
| Refractive Index | 1.527 |
| Quantity | 2500 g |
| UN Number | UN1993 |
| Beilstein | 774102 |
| Solubility Information | Soluble in water(0.14g/L). |
| Show More |
Safety and Handling
| Hazard Category | H226-H312-H315-H319-H332-H335 |
| Hazard Statement | GHS H Statement H226-H332-H319 Flammable liquid and vapor. Harmful if inhaled. Causes serious eye irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 207-300-2 |
| RTECSNumber | CY9033750 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only