missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-fluoro-2-iodobenzene 98.0+%, TCI America™

Catalog No. p-7123583
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45065G 5 g
B450625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45065G Supplier TCI America Supplier No. B45065G
Only null left

Chemical Identifiers

CAS 202865-72-3
Molecular Formula C6H3BrFI
Molecular Weight (g/mol) 300.897
MDL Number MFCD00042578
InChI Key QDCBSSUCEUMLGC-UHFFFAOYSA-N
Synonym 2-bromo-5-fluoroiodobenzene, 4-fluoro-2-iodobromobenzene, 2-iodo-4-fluorobromobenzene, 1-bromo-4-fluoro-2-iodo-benzene, benzene, 1-bromo-4-fluoro-2-iodo, 2-bromo-5-fluoro-1-iodobenzene, pubchem3233, acmc-1cljm, ksc493k3j, 1-bromo-2-iodo-4-fluorobenzene
PubChem CID 2773370
IUPAC Name 1-bromo-4-fluoro-2-iodobenzene
SMILES C1=CC(=C(C=C1F)I)Br

Specifications

Color Yellow
Boiling Point 235°C
Quantity 5 g
Formula Weight 300.90
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-fluoro-2-iodobenzene