missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-fluoro-2-iodobenzene, 98+%

Catalog No. p-7028143
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2528906 5 g
AAB2528914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2528906 Supplier Thermo Scientific Chemicals Supplier No. B2528906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 202865-72-3 | C6H3BrFI | 300.897 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 202865-72-3
Molecular Formula C6H3BrFI
Molecular Weight (g/mol) 300.897
MDL Number MFCD00042578
InChI Key QDCBSSUCEUMLGC-UHFFFAOYSA-N
Synonym 2-bromo-5-fluoroiodobenzene, 4-fluoro-2-iodobromobenzene, 2-iodo-4-fluorobromobenzene, 1-bromo-4-fluoro-2-iodo-benzene, benzene, 1-bromo-4-fluoro-2-iodo, 2-bromo-5-fluoro-1-iodobenzene, pubchem3233, acmc-1cljm, ksc493k3j, 1-bromo-2-iodo-4-fluorobenzene
PubChem CID 2773370
IUPAC Name 1-bromo-4-fluoro-2-iodobenzene
SMILES C1=CC(=C(C=C1F)I)Br

Specifications

Density 2.3
Boiling Point 235°C
Flash Point >110°C (230°F)
Refractive Index 1.63
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 300.9
Percent Purity ≥98%
Chemical Name or Material 1-Bromo-4-fluoro-2-iodobenzene
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only