missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-(ethylthio)benzene, 97%

Catalog No. AAL1371822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1371822 100 g
AAL1371806 5 g
AAL1371814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1371822 Supplier Thermo Scientific Chemicals Supplier No. L1371822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 30506-30-0 | C8H9BrS | 217.124 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30506-30-0
Molecular Formula C8H9BrS
Molecular Weight (g/mol) 217.124
MDL Number MFCD01318109
InChI Key CTFHVAKXUMMGOC-UHFFFAOYSA-N
Synonym 1-bromo-4-ethylthio benzene, 1-bromo-4-ethylsulfanyl benzene, 4-bromophenyl ethyl sulfane, benzene,1-bromo-4-ethylthio, acmc-1ae1n, 1-bromo-4-ethylthiobenzene, 4-bromo-1-ethylthiobenzene, ethyl 4-bromophenyl sulfide, 1-bromo-4-ethylthio-benzene, 1-bromo-4-ethylsulfanyl-benzene
PubChem CID 7006650
IUPAC Name 1-bromo-4-ethylsulfanylbenzene
SMILES CCSC1=CC=C(C=C1)Br

Specifications

Boiling Point 109°C to 111°C (2 mmHg)
Refractive Index 1.6069
Quantity 100 g
Beilstein 2354970
Formula Weight 217.13
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-(ethylthio)benzene
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only