missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-(dimethylamino)naphthalene, 95%

Catalog No. AAL1766603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1766603 1 g
AAL1766606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1766603 Supplier Thermo Scientific Chemicals Supplier No. L1766603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 59557-93-6 | C12H12BrN | 250.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59557-93-6
Molecular Formula C12H12BrN
Molecular Weight (g/mol) 250.139
MDL Number MFCD02093945
InChI Key CWPDFSZHOZTGQW-UHFFFAOYSA-N
Synonym 1-bromo-4-dimethylamino naphthalene, acmc-20aox1, 4-bromonaphthyl dimethylamine, n,n-dimethyl-4-bromo-1-naphthylamine, n,n-dimethyl-4-bromonaphthalene-1-amine
PubChem CID 4083804
IUPAC Name 4-bromo-N,N-dimethylnaphthalen-1-amine
SMILES CN(C)C1=CC=C(C2=CC=CC=C21)Br

Specifications

Density 1.4305
Boiling Point 137°C to 139°C (2 mmHg)
Refractive Index 1.6361
Quantity 1 g
UN Number UN2810
Beilstein 3254839
Formula Weight 250.14
Percent Purity 95%
Chemical Name or Material 1-Bromo-4-(dimethylamino)naphthalene
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only