accessibility menu, dialog, popup

UserName

CAS RN 59557-93-6

IUPAC Name: 4-bromo-N,N-dimethylnaphthalen-1-amine
Synonyms: 4-bromo-N,N-dimethylnaphthalen-1-amine
Molecular Weight (g/mol): 250.14
Molecular Formula: C12H12BrN
InChi Key: CWPDFSZHOZTGQW-UHFFFAOYSA-N
SMILES: CN(C)C1=C2C=CC=CC2=C(Br)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)