missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-(difluoromethoxy)benzene, 97%

Catalog No. AAB2010606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2010606 5 g
AAB2010614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2010606 Supplier Thermo Scientific Chemicals Supplier No. B2010606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5905-69-1 | C7H5BrF2O | 223.017 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5905-69-1
Molecular Formula C7H5BrF2O
Molecular Weight (g/mol) 223.017
MDL Number MFCD00221465
InChI Key ORIYZUFTROJBQJ-UHFFFAOYSA-N
Synonym 1-bromo-4-difluoromethoxy benzene, 4-difluoromethoxy bromobenzene, p-difluoromethoxybromobenzene, 1-difluoromethoxy-4-bromobenzene, benzene, 1-bromo-4-difluoromethoxy, l-bromo-4-difluoromethoxy benzene, pubchem4309, acmc-209m9x, p-difluoro-methoxybromobenzene, ksc495s1h
PubChem CID 2737007
IUPAC Name 1-bromo-4-(difluoromethoxy)benzene
SMILES C1=CC(=CC=C1OC(F)F)Br

Specifications

Density 1.631
Boiling Point 96°C to 98°C (15 mmHg)
Refractive Index 1.501
Quantity 5 g
UN Number UN1993
Beilstein 2521410
Formula Weight 223.02
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-(difluoromethoxy)benzene
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only