missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-decylbenzene 88.0+%, TCI America™

Catalog No. p-7119156
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16111G 1 g
B16115G 5 g
B161125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B16111G Supplier TCI America Supplier No. B16111G
Only null left

Chemical Identifiers

CAS 106418-67-1
Molecular Formula C16H25Br
Molecular Weight (g/mol) 297.28
MDL Number MFCD00061112
InChI Key SWTGVSCSSDYINB-UHFFFAOYSA-N
Synonym 1-bromo-4-n-decylbenzene, benzene, 1-bromo-4-decyl, 4-decylbromobenzene, acmc-2098jm, ksc909c2n
PubChem CID 13757384
IUPAC Name 1-bromo-4-decylbenzene
SMILES CCCCCCCCCCC1=CC=C(Br)C=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 297.28
Percent Purity ≥88.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-decylbenzene