missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-chlorobenzene 99.0+%, TCI America™

Catalog No. p-7118534
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B057125G 25 g
B0571100G 100 g
B0571500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B057125G Supplier TCI America Supplier No. B057125G
Only null left

Chemical Identifiers

CAS 106-39-8
Molecular Formula C6H4BrCl
Molecular Weight (g/mol) 191.45
MDL Number MFCD00000600
InChI Key NHDODQWIKUYWMW-UHFFFAOYSA-N
Synonym 4-bromochlorobenzene, p-bromochlorobenzene, 4-chlorobromobenzene, p-chlorobromobenzene, p-chlorophenyl bromide, benzene, 1-bromo-4-chloro, 4-chlorophenyl bromide, 1-chloro-4-bromobenzene, 4-chloro-1-bromobenzene, p-bromoclorobenzene
PubChem CID 7806
IUPAC Name 1-bromo-4-chlorobenzene
SMILES ClC1=CC=C(Br)C=C1

Specifications

Melting Point 67°C
Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Boiling Point 196°C
Formula Weight 191.45
Percent Purity ≥99.0% (GC)
Chemical Name or Material 1-Bromo-4-chlorobenzene
EINECSNumber (3)-0066&(3)-0070
RTECSNumber CY9006000
TSCA Yes