accessibility menu, dialog, popup

UserName

CAS RN 106-39-8

IUPAC Name: 1-bromo-4-chlorobenzene
Synonyms: bromochlorobenzene P-bromochlorobenzene
Molecular Weight (g/mol): 191.45
Molecular Formula: C6H4BrCl
InChi Key: NHDODQWIKUYWMW-UHFFFAOYSA-N
SMILES: ClC1=CC=C(Br)C=C1
Percent Purity 
  • (2)
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (6)
  • (2)
  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)