missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-chloro-2-nitrobenzene, 98%

Catalog No. AAA1263506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1263506 5 g
AAA1263514 25 g
AAA1263522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1263506 Supplier Thermo Scientific Chemicals Supplier No. A1263506

CAS: 41513-04-6 | C6H3BrClNO2 | 236.449 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 41513-04-6
Molecular Formula C6H3BrClNO2
Molecular Weight (g/mol) 236.449
MDL Number MFCD00024320
InChI Key UKTIMFAJRPSNGR-UHFFFAOYSA-N
Synonym 2-bromo-5-chloronitrobenzene, benzene, 1-bromo-4-chloro-2-nitro, 4-bromo-3-nitrochlorobenzene, 3-chloro-6-bromonitrobenzene, pubchem3593, acmc-209jkn, 1-bromo-4-chloronitrobenzene, ksc490m7r, 2-bromo-5-chloro-nitrobenzene, bromo-4-chloro-2-nitrobenzene
PubChem CID 2794904
IUPAC Name 1-bromo-4-chloro-2-nitrobenzene
SMILES C1=CC(=C(C=C1Cl)[N+](=O)[O-])Br

Specifications

Melting Point 68°C to 72°C
Quantity 5 g
Beilstein 1950424
Formula Weight 236.45
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-chloro-2-nitrobenzene
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 255-421-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only