missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-4-butylbenzene 97.0+%, TCI America™
Supplier: TCI America B190725G
Specifications
| 1-Bromo-4-butylbenzene | |
| Yellow | |
| C10H13Br | |
| 25 g | |
| BRGVKVZXDWGJBX-UHFFFAOYSA-N | |
| 1-bromo-4-butylbenzene | |
| 521059 | |
| ≥97.0% (GC) |
| 41492-05-1 | |
| 246°C | |
| MFCD00040934 | |
| 1-bromo-4-n-butylbenzene, 4-bromo-n-butylbenzene, 4-n-butylbromobenzene, 1-4-bromophenyl butane, benzene, 1-bromo-4-butyl, 4-butyl bromobenzene, 4-butyl-bromobenzene, 4-1-butylbromobenzene, 4-butyl phenylbromide, pubchem15109 | |
| CCCCC1=CC=C(C=C1)Br | |
| 213.118 | |
| 213.12 | |
| Liquid |
Chemical Identifiers
| 41492-05-1 | |
| 213.118 | |
| BRGVKVZXDWGJBX-UHFFFAOYSA-N | |
| 521059 | |
| CCCCC1=CC=C(C=C1)Br |
| C10H13Br | |
| MFCD00040934 | |
| 1-bromo-4-n-butylbenzene, 4-bromo-n-butylbenzene, 4-n-butylbromobenzene, 1-4-bromophenyl butane, benzene, 1-bromo-4-butyl, 4-butyl bromobenzene, 4-butyl-bromobenzene, 4-1-butylbromobenzene, 4-butyl phenylbromide, pubchem15109 | |
| 1-bromo-4-butylbenzene |
Safety and Handling
TSCA : No