missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-butylbenzene 97.0+%, TCI America™

Catalog No. B19075G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19075G 5 g
B190725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19075G Supplier TCI America Supplier No. B19075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41492-05-1
Molecular Formula C10H13Br
Molecular Weight (g/mol) 213.118
MDL Number MFCD00040934
InChI Key BRGVKVZXDWGJBX-UHFFFAOYSA-N
Synonym 1-bromo-4-n-butylbenzene, 4-bromo-n-butylbenzene, 4-n-butylbromobenzene, 1-4-bromophenyl butane, benzene, 1-bromo-4-butyl, 4-butyl bromobenzene, 4-butyl-bromobenzene, 4-1-butylbromobenzene, 4-butyl phenylbromide, pubchem15109
PubChem CID 521059
IUPAC Name 1-bromo-4-butylbenzene
SMILES CCCCC1=CC=C(C=C1)Br

Specifications

Color Yellow
Boiling Point 246°C
Quantity 5 g
Formula Weight 213.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-butylbenzene