missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4,6-nonadecadiyne 96.0+%, TCI America™

Catalog No. B1342100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1342100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1342100MG Supplier TCI America Supplier No. B1342100MG
Only null left

Chemical Identifiers

CAS 376591-04-7
Molecular Formula C19H31Br
Molecular Weight (g/mol) 339.361
MDL Number MFCD00059031
InChI Key BCWJEWJRRDCQQU-UHFFFAOYSA-N
Synonym 4,6-Nonadecadiynyl Bromide
PubChem CID 44630135
IUPAC Name 1-bromononadeca-4,6-diyne
SMILES CCCCCCCCCCCCC#CC#CCCCBr

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 339.36
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4,6-nonadecadiyne
TSCA No
Recommended Storage Freezer