missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-(1,1-difluoroethyl)benzene, 95%

Catalog No. AC432530100
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432530100 10 g Glass bottle
AC432530050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432530100 Supplier Thermo Scientific Chemicals Supplier No. 432530100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1000994-95-5 | C8H7BrF2 | 221.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1000994-95-5
Molecular Formula C8H7BrF2
Molecular Weight (g/mol) 221.05
MDL Number MFCD11110333
InChI Key QXIBKCFAFRHORF-UHFFFAOYSA-N
Synonym 1-bromo-4-1,1-difluoroethyl benzene, 1-bromo-4-1,1-difluoro-ethyl-benzene, benzene, 1-bromo-4-1,1-difluoroethyl, 4-1,1-difluoroethyl bromobenzene, bromo-4-1,1-difluoroethyl benzene, 1-bromo-4-1,1-difluoroethyl-benzene, 1-4-bromophenyl-1,1-difluoroethane
PubChem CID 45158847
IUPAC Name 1-bromo-4-(1,1-difluoroethyl)benzene
SMILES CC(C1=CC=C(C=C1)Br)(F)F

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Brown
Boiling Point 80.0°C to 82.0°C (10.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 10 g
Formula Weight 221.05
Percent Purity 95%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 1-Bromo-4-(1, 1-difluoroethyl)benzene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only