missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-3-(trifluoromethoxy)benzene 98.0+%, TCI America™
$55.92 - $111.84
Chemical Identifiers
| CAS | 2252-44-0 |
|---|---|
| Molecular Formula | C7H4BrF3O |
| Molecular Weight (g/mol) | 241.007 |
| MDL Number | MFCD00040943 |
| InChI Key | WVUDHWBCPSXAFN-UHFFFAOYSA-N |
| Synonym | 1-bromo-3-trifluoromethoxy benzene, 3-trifluoromethoxy bromobenzene, 3-bromo trifluoromethoxy benzene, m-bromo trifluoromethoxy benzene, 3-bromo-1-trifluoromethoxy benzene, benzene, 1-bromo-3-trifluoromethoxy, 3-bromo-alpha,alpha,alpha-trifluoroanisole, bromo-3-trifluoromethoxy benzene, buttpark 91\57-27, 1-bromo-3-trifluoromethoxy-benzene |
| PubChem CID | 519964 |
| IUPAC Name | 1-bromo-3-(trifluoromethoxy)benzene |
| SMILES | C1=CC(=CC(=C1)Br)OC(F)(F)F |
Chemical Identifiers
| 2252-44-0 | |
| 241.007 | |
| WVUDHWBCPSXAFN-UHFFFAOYSA-N | |
| 519964 | |
| C1=CC(=CC(=C1)Br)OC(F)(F)F |
| C7H4BrF3O | |
| MFCD00040943 | |
| 1-bromo-3-trifluoromethoxy benzene, 3-trifluoromethoxy bromobenzene, 3-bromo trifluoromethoxy benzene, m-bromo trifluoromethoxy benzene, 3-bromo-1-trifluoromethoxy benzene, benzene, 1-bromo-3-trifluoromethoxy, 3-bromo-alpha,alpha,alpha-trifluoroanisole, bromo-3-trifluoromethoxy benzene, buttpark 91\57-27, 1-bromo-3-trifluoromethoxy-benzene | |
| 1-bromo-3-(trifluoromethoxy)benzene |
Specifications
| 2252-44-0 | |
| 81°C | |
| MFCD00040943 | |
| 1-bromo-3-trifluoromethoxy benzene, 3-trifluoromethoxy bromobenzene, 3-bromo trifluoromethoxy benzene, m-bromo trifluoromethoxy benzene, 3-bromo-1-trifluoromethoxy benzene, benzene, 1-bromo-3-trifluoromethoxy, 3-bromo-alpha,alpha,alpha-trifluoroanisole, bromo-3-trifluoromethoxy benzene, buttpark 91\57-27, 1-bromo-3-trifluoromethoxy-benzene | |
| C1=CC(=CC(=C1)Br)OC(F)(F)F | |
| 241.007 | |
| 241.01 | |
| Liquid |
| Yellow | |
| C7H4BrF3O | |
| 5 g | |
| WVUDHWBCPSXAFN-UHFFFAOYSA-N | |
| 1-bromo-3-(trifluoromethoxy)benzene | |
| 519964 | |
| ≥98.0% (GC) | |
| 1-Bromo-3-(trifluoromethoxy)benzene |
Safety and Handling
TSCA : No