missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-(trifluoromethoxy)benzene 98.0+%, TCI America™

Catalog No. B20075G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20075G 5 g
B200725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20075G Supplier TCI America Supplier No. B20075G

Chemical Identifiers

CAS 2252-44-0
Molecular Formula C7H4BrF3O
Molecular Weight (g/mol) 241.007
MDL Number MFCD00040943
InChI Key WVUDHWBCPSXAFN-UHFFFAOYSA-N
Synonym 1-bromo-3-trifluoromethoxy benzene, 3-trifluoromethoxy bromobenzene, 3-bromo trifluoromethoxy benzene, m-bromo trifluoromethoxy benzene, 3-bromo-1-trifluoromethoxy benzene, benzene, 1-bromo-3-trifluoromethoxy, 3-bromo-alpha,alpha,alpha-trifluoroanisole, bromo-3-trifluoromethoxy benzene, buttpark 91\57-27, 1-bromo-3-trifluoromethoxy-benzene
PubChem CID 519964
IUPAC Name 1-bromo-3-(trifluoromethoxy)benzene
SMILES C1=CC(=CC(=C1)Br)OC(F)(F)F

Specifications

Color Yellow
Boiling Point 81°C
Quantity 5 g
Formula Weight 241.01
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-3-(trifluoromethoxy)benzene