missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-(trifluoromethoxy)benzene, 98+%, Thermo Scientific Chemicals

$263.03 - $263.03

Chemical Identifiers

CAS 2252-44-0
Molecular Formula C7H4BrF3O
Molecular Weight (g/mol) 241.007
MDL Number MFCD00040943
InChI Key WVUDHWBCPSXAFN-UHFFFAOYSA-N
Synonym 1-bromo-3-trifluoromethoxy benzene, 3-trifluoromethoxy bromobenzene, 3-bromo trifluoromethoxy benzene, m-bromo trifluoromethoxy benzene, 3-bromo-1-trifluoromethoxy benzene, benzene, 1-bromo-3-trifluoromethoxy, 3-bromo-alpha,alpha,alpha-trifluoroanisole, bromo-3-trifluoromethoxy benzene, buttpark 91\57-27, 1-bromo-3-trifluoromethoxy-benzene
PubChem CID 519964
IUPAC Name 1-bromo-3-(trifluoromethoxy)benzene
SMILES C1=CC(=CC(=C1)Br)OC(F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1656114
View Documents
Thermo Scientific Chemicals
A1656114
25 g
Each for $263.03
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2252-44-0
241.007
WVUDHWBCPSXAFN-UHFFFAOYSA-N
519964
C1=CC(=CC(=C1)Br)OC(F)(F)F
C7H4BrF3O
MFCD00040943
1-bromo-3-trifluoromethoxy benzene, 3-trifluoromethoxy bromobenzene, 3-bromo trifluoromethoxy benzene, m-bromo trifluoromethoxy benzene, 3-bromo-1-trifluoromethoxy benzene, benzene, 1-bromo-3-trifluoromethoxy, 3-bromo-alpha,alpha,alpha-trifluoroanisole, bromo-3-trifluoromethoxy benzene, buttpark 91\57-27, 1-bromo-3-trifluoromethoxy-benzene
1-bromo-3-(trifluoromethoxy)benzene

Specifications

2252-44-0
156°C to 158°C
C7H4BrF3O
MFCD00040943
1945938
WVUDHWBCPSXAFN-UHFFFAOYSA-N
1-bromo-3-(trifluoromethoxy)benzene
519964
≥98%
1.62
62°C (143°F)
1.462
25 g
1-bromo-3-trifluoromethoxy benzene, 3-trifluoromethoxy bromobenzene, 3-bromo trifluoromethoxy benzene, m-bromo trifluoromethoxy benzene, 3-bromo-1-trifluoromethoxy benzene, benzene, 1-bromo-3-trifluoromethoxy, 3-bromo-alpha,alpha,alpha-trifluoroanisole, bromo-3-trifluoromethoxy benzene, buttpark 91\57-27, 1-bromo-3-trifluoromethoxy-benzene
C1=CC(=CC(=C1)Br)OC(F)(F)F
241.007
241.01
1-Bromo-3-(trifluoromethoxy)benzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only