missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-(trifluoromethoxy)benzene, 98+%, Thermo Scientific Chemicals

Catalog No. AAA1656114
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1656114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1656114 Supplier Thermo Scientific Chemicals Supplier No. A1656114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2252-44-0
Molecular Formula C7H4BrF3O
Molecular Weight (g/mol) 241.007
MDL Number MFCD00040943
InChI Key WVUDHWBCPSXAFN-UHFFFAOYSA-N
Synonym 1-bromo-3-trifluoromethoxy benzene, 3-trifluoromethoxy bromobenzene, 3-bromo trifluoromethoxy benzene, m-bromo trifluoromethoxy benzene, 3-bromo-1-trifluoromethoxy benzene, benzene, 1-bromo-3-trifluoromethoxy, 3-bromo-alpha,alpha,alpha-trifluoroanisole, bromo-3-trifluoromethoxy benzene, buttpark 91\57-27, 1-bromo-3-trifluoromethoxy-benzene
PubChem CID 519964
IUPAC Name 1-bromo-3-(trifluoromethoxy)benzene
SMILES C1=CC(=CC(=C1)Br)OC(F)(F)F

Specifications

Density 1.62
Boiling Point 156°C to 158°C
Flash Point 62°C (143°F)
Refractive Index 1.462
Quantity 25 g
Beilstein 1945938
Formula Weight 241.01
Percent Purity ≥98%
Chemical Name or Material 1-Bromo-3-(trifluoromethoxy)benzene
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only