missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-phenylpropane, 98%

Catalog No. AAA1405022
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1405022 100 g
AAA1405036 500 g
AAA140500E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1405022 Supplier Thermo Scientific Chemicals Supplier No. A1405022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 637-59-2 | C9H11Br | 199.09 g/mol

1-Bromo-3-phenylpropane is used in the synthesis of MraY natural inhibitors as antibacterial agents. Also used in the preparation of quinolinone derivatives as potent and selective MAO-B inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3-phenylpropane is used in the synthesis of MraY natural inhibitors as antibacterial agents. Also used in the preparation of quinolinone derivatives as potent and selective MAO-B inhibitors.

Solubility
Soluble in water (partly). Soluble in chloroform and methanol.

Notes
Store in cool, dry place in tightly closed containers. Store away from oxidizing agent.

Chemical Identifiers

CAS 637-59-2
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.09
MDL Number MFCD00000257
InChI Key XMZQWZJMTBCUFT-UHFFFAOYSA-N
Synonym 3-bromopropyl benzene, 1-bromo-3-phenylpropane, 3-phenylpropyl bromide, benzene, 3-bromopropyl, 3-bromo-1-phenylpropane, 3-bromoprop-1-yl benzene, unii-6vx623qn9v, .gamma.-bromopropyl benzene, 4-bromopropylbenzene, benzene, bromopropyl
PubChem CID 12503
IUPAC Name 3-bromopropylbenzene
SMILES BrCCCC1=CC=CC=C1

Specifications

Density 1.308
Boiling Point 219°C to 220°C
Flash Point 101°C (213°F)
Refractive Index 1.546
Quantity 100 g
Beilstein 2205527
Solubility Information Soluble in water (partly). Soluble in chloroform and methanol.
Formula Weight 199.09
Percent Purity 98%
Chemical Name or Material 1-Bromo-3-phenylpropane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-294-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only